siRNA Details
| Gene Symbol: FAM186A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145475.1 | NP_001138947.1 | 4 | 5183 | ||
| XM_006719231.3 | XP_006719294.1 | 4 | 5194 | |||
| XM_011537890.2 | XP_011536192.1 | 4 | 5194 | |||
| XM_011537892.1 | XP_011536194.1 | 3 | 4703 | |||

FAM186A
Genomic Map