siRNA Details
| Gene Symbol: CCDC168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146197.1 | NP_001139669.1 | 4 | 6233 | ||
| XM_011521106.1 | XP_011519408.1 | 5 | 6106 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC168| Gene Symbol: CCDC168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146197.1 | NP_001139669.1 | 4 | 6233 | ||
| XM_011521106.1 | XP_011519408.1 | 5 | 6106 | |||