siRNA Details
| Gene Symbol: KLHL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109997.2 | NP_001103467.2 | 4 | 1521 | ||
| XM_005267347.4 | XP_005267404.1 | 5 | 2520 | |||
| XM_011536450.2 | XP_011534752.1 | 5 | 2236 | |||
| GenBank mRNA | BC146951.1 | - | 3 | 1115 | 9056879 | |
| BC146954.1 | - | 3 | 1115 | 9056882 | ||
| BC151130.1 | - | 3 | 1137 | 9057016 | ||

KLHL33
Genomic Map