siRNA Details
| Gene Symbol: GSE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005255859.4 | XP_005255916.3 | 1 | 945 | ||
| XM_005255860.3 | XP_005255917.3 | 1 | 952 | |||
| XM_005255861.4 | XP_005255918.3 | 1 | 945 | |||
| XM_017023084.1 | XP_016878573.1 | 1 | 1117 | |||

GSE1
Genomic Map