siRNA Details
| Gene Symbol: IQCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160042.1 | NP_001153514.1 | 3 | 512 | ||
| NM_018134.2 | NP_060604.2 | 3 | 272 | |||
| GenBank mRNA | AK001409.1 | - | 3 | 236 | ||
| AK223272.1 | - | 3 | 260 | |||
| AK299235.1 | - | 2 | 447 | |||
| AK307853.1 | - | 3 | 476 | |||
| BC098250.1 | - | 3 | 235 | |||
| BC098260.1 | - | 3 | 235 | 40024457 | ||
| BC098317.1 | - | 3 | 235 | 40024460 | ||
| BC098366.1 | - | 3 | 235 | 40024462 | ||
| DA746781.1 | - | 3 | 272 | |||

IQCC
Genomic Map