siRNA Details
| Gene Symbol: C1orf194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122961.1 | NP_001116433.1 | 5 | 658 | ||
| XM_005270448.3 | XP_005270505.1 | 4 | 561 | |||
| XM_006710342.3 | XP_006710405.1 | 5 | 1004 | |||
| XM_006710343.3 | XP_006710406.1 | 4 | 887 | |||
| XM_011540647.2 | XP_011538949.1 | 5 | 1035 | |||
| GenBank mRNA | BC127905.1 | - | 3 | 349 | ||
| BG196327.1 | - | 3 | 321 | |||

C1orf194
Genomic Map