siRNA Details
| Gene Symbol: OCSTAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080721.2 | NP_542452.1 | 2 | 775 | ||
| XM_011528532.2 | XP_011526834.1 | 1 | 585 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OCSTAMP| Gene Symbol: OCSTAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080721.2 | NP_542452.1 | 2 | 775 | ||
| XM_011528532.2 | XP_011526834.1 | 1 | 585 | |||