siRNA Details
| Gene Symbol: TRABD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277053.1 | NP_001263982.1 | 3, 4 | 1023 | ||
| NM_001307978.1 | NP_001294907.1 | 3, 4 | 1023 | |||
| XM_011532504.1 | XP_011530806.1 | 3, 4 | 823 | |||
| GenBank mRNA | AK296608.1 | - | 3, 4 | 860 | ||
| AK309615.1 | - | 3, 4 | 862 | |||
| BC049209.1 | - | 2, 3 | 563 | |||
| JQ653415.1 | - | 3, 4 | 817 | |||

TRABD2A
Genomic Map