siRNA Details
Gene Symbol: UBE2QL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145161.2 | NP_001138633.1 | 2 | 3584 | ||
GenBank mRNA | AK096612.1 | - | 1 | 2618 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: UBE2QL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145161.2 | NP_001138633.1 | 2 | 3584 | ||
GenBank mRNA | AK096612.1 | - | 1 | 2618 |