siRNA Details
| Gene Symbol: ZFP92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136273.1 | NP_001129745.1 | 2 | 95 | ||
| XM_005274652.3 | XP_005274709.1 | 4 | 405 | |||
| XM_011531115.2 | XP_011529417.1 | 3 | 356 | |||
| XM_017029281.1 | XP_016884770.1 | 5 | 754 | |||

ZFP92
Genomic Map