siRNA Details
| Gene Symbol: ISM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080826.1 | NP_543016.1 | 2 | 158 | ||
| XM_017027680.1 | XP_016883169.1 | 2 | 158 | |||
| GenBank mRNA | DB169194.1 | - | 2 | 160 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ISM1| Gene Symbol: ISM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080826.1 | NP_543016.1 | 2 | 158 | ||
| XM_017027680.1 | XP_016883169.1 | 2 | 158 | |||
| GenBank mRNA | DB169194.1 | - | 2 | 160 | ||