siRNA Details
| Gene Symbol: DNAJC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018163.2 | NP_060633.1 | 3, 4 | 246 | ||
| XM_017022401.1 | XP_016877890.1 | 4, 5 | 621 | |||
| XM_017022402.1 | XP_016877891.1 | 3, 4 | 292 | |||
| GenBank mRNA | AK001496.1 | - | 3, 4 | 225 | ||
| AK308659.1 | - | 3, 4 | 225 | |||
| AK308858.1 | - | 3, 4 | 241 | |||
| AK309307.1 | - | 3, 4 | 242 | |||
| AK310140.1 | - | 3, 4 | 245 | |||
| BC000048.2 | - | 3, 4 | 232 | 3507470 | ||
| DA341962.1 | - | 3, 4 | 246 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_139139, AK077385, BC013487, BY794890.

DNAJC17
Genomic Map