siRNA Details
| Gene Symbol: TMEM120B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080825.2 | NP_001074294.2 | 12 | 1670 | ||
| XM_005253843.4 | XP_005253900.1 | 8 | 1778 | |||
| XM_017018853.1 | XP_016874342.1 | 12 | 1670 | |||
| XM_017018854.1 | XP_016874343.1 | 10 | 1542 | |||
| GenBank mRNA | AK126568.1 | - | 12 | 1670 | ||
| BC035884.1 | - | 11 | 1422 | |||
| BX648084.1 | - | 8 | 1143 | |||

TMEM120B
Genomic Map