siRNA Details
| Gene Symbol: TSPAN19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100917.1 | NP_001094387.1 | 8, 9 | 787 | ||
| XM_005268669.4 | XP_005268726.1 | 8, 9 | 837 | |||
| XM_017018862.1 | XP_016874351.1 | 7, 8 | 725 | |||
| XM_017018863.1 | XP_016874352.1 | 7, 8 | 719 | |||

TSPAN19
Genomic Map