siRNA Details
| Gene Symbol: LINC00410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027039.1 | 3, 4 | 366 | |||
| GenBank mRNA | AK055781.1 | - | 3, 4 | 366 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00410| Gene Symbol: LINC00410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027039.1 | 3, 4 | 366 | |||
| GenBank mRNA | AK055781.1 | - | 3, 4 | 366 | ||