siRNA Details
| Gene Symbol: LINC00343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046391.2 | 2 | 559 | |||
| NR_120418.1 | 2 | 559 | ||||
| GenBank mRNA | AK058053.1 | - | 2 | 520 | ||
| BC126908.1 | - | 2 | 498 | |||
| BC127249.1 | - | 2 | 498 | |||
| CD244335.1 | - | 2 | 538 | |||

LINC00343
Genomic Map