siRNA Details
| Gene Symbol: LINC00637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038436.1 | 3 | 1669 | |||
| GenBank mRNA | AK057986.1 | - | 3 | 1671 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00637| Gene Symbol: LINC00637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038436.1 | 3 | 1669 | |||
| GenBank mRNA | AK057986.1 | - | 3 | 1671 | ||