siRNA Details
| Gene Symbol: C15orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198784.1 | NP_001185713.1 | 2 | 434 | ||
| GenBank mRNA | AK130862.1 | - | 2 | 411 | ||
| BX648577.1 | - | 2 | 289 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C15orf65| Gene Symbol: C15orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198784.1 | NP_001185713.1 | 2 | 434 | ||
| GenBank mRNA | AK130862.1 | - | 2 | 411 | ||
| BX648577.1 | - | 2 | 289 | |||