siRNA Details
| Gene Symbol: C15orf61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143936.1 | NP_001137408.1 | 2 | 497 | ||
| GenBank mRNA | AK294012.1 | - | 2 | 432 | ||
| AY726587.1 | - | 2 | 389 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C15orf61| Gene Symbol: C15orf61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143936.1 | NP_001137408.1 | 2 | 497 | ||
| GenBank mRNA | AK294012.1 | - | 2 | 432 | ||
| AY726587.1 | - | 2 | 389 | |||