siRNA Details
| Gene Symbol: SLC38A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080442.2 | NP_001073911.1 | 8 | 1137 | ||
| XM_017022946.1 | XP_016878435.1 | 10 | 1602 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC38A8| Gene Symbol: SLC38A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080442.2 | NP_001073911.1 | 8 | 1137 | ||
| XM_017022946.1 | XP_016878435.1 | 10 | 1602 | |||