siRNA Details
| Gene Symbol: C16orf92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109659.1 | NP_001103129.1 | 4 | 413 | ||
| NM_001109660.1 | NP_001103130.1 | 4 | 493 | |||
| GenBank mRNA | AK058133.1 | - | 4 | 493 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16orf92| Gene Symbol: C16orf92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109659.1 | NP_001103129.1 | 4 | 413 | ||
| NM_001109660.1 | NP_001103130.1 | 4 | 493 | |||
| GenBank mRNA | AK058133.1 | - | 4 | 493 | ||