siRNA Details
| Gene Symbol: IGSF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205280.1 | NP_001192209.1 | 2, 3 | 401 | ||
| XM_017026317.1 | XP_016881806.1 | 2, 3 | 575 | |||
| XM_017026318.1 | XP_016881807.1 | 2, 3 | 1454 | |||
| XM_017026319.1 | XP_016881808.1 | 2, 3 | 575 | |||
| XM_017026320.1 | XP_016881809.1 | 3, 4 | 656 | |||
| XM_017026321.1 | XP_016881810.1 | 3, 4 | 683 | |||
| XM_017026322.1 | XP_016881811.1 | 3, 4 | 962 | |||
| XM_017026323.1 | XP_016881812.1 | 2, 3 | 575 | |||
| GenBank mRNA | BC127928.1 | - | 1, 2 | 221 | ||
| BC127929.1 | - | 1, 2 | 221 | |||
| BX112065.1 | - | 2, 3 | 401 | |||

IGSF23
Genomic Map