siRNA Details
| Gene Symbol: OVAAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125716.1 | 3 | 843 | |||
| GenBank mRNA | BC036830.1 | - | 3 | 840 | ||
| BC044260.1 | - | 3 | 841 | |||
| BC063624.1 | - | 3 | 843 | |||
| BC073892.1 | - | 3 | 867 | |||
| BC107121.2 | - | 2 | 747 | |||
| BC107122.1 | - | 2 | 747 | |||

OVAAL
Genomic Map