siRNA Details
| Gene Symbol: LOC105378853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135039.1 | 1 | 109 | |||
| GenBank mRNA | BX103107.1 | - | 1 | 109 | ||
| Gene Symbol: LOC149351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022898.1 | - | 2 | 373 | ||
| BC036441.1 | - | 2 | 578 | |||

LOC105378853
Genomic Map