siRNA Details
| Gene Symbol: LINC01315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120595.1 | 2 | 1067 | |||
| NR_120596.1 | 2 | 648 | ||||
| NR_120597.1 | 2 | 305 | ||||
| GenBank mRNA | AV709904.1 | - | 2 | 639 | ||
| BC038245.1 | - | 2 | 1033 | |||
| CB050274.1 | - | 2 | 314 | |||

LINC01315
Genomic Map