siRNA Details
| Gene Symbol: LINC01102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015399.1 | 4 | 989 | |||
| GenBank mRNA | AL137513.1 | - | 4 | 958 | ||
| BC105699.1 | - | 4 | 883 | |||
| BC105700.1 | - | 4 | 883 | |||
| BC105701.1 | - | 4 | 883 | |||
| BC105702.1 | - | 4 | 883 | |||

LINC01102
Genomic Map