siRNA Details
| Gene Symbol: MIR3681HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110197.1 | 4 | 558 | |||
| GenBank mRNA | BX439624.2 | - | 4 | 562 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR3681HG| Gene Symbol: MIR3681HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110197.1 | 4 | 558 | |||
| GenBank mRNA | BX439624.2 | - | 4 | 562 | ||