siRNA Details
| Gene Symbol: LINC00608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015390.1 | 2 | 815 | |||
| NR_024385.1 | 2 | 1291 | ||||
| GenBank mRNA | AK098539.1 | - | 2 | 1291 | ||
| BC033552.1 | - | 2 | 1273 | |||
| BC049373.1 | - | 2 | 815 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00608| Gene Symbol: LINC00608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015390.1 | 2 | 815 | |||
| NR_024385.1 | 2 | 1291 | ||||
| GenBank mRNA | AK098539.1 | - | 2 | 1291 | ||
| BC033552.1 | - | 2 | 1273 | |||
| BC049373.1 | - | 2 | 815 | |||