siRNA Details
| Gene Symbol: LOC101928327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130782.1 | 1 | 172 | |||
| GenBank mRNA | BC021184.1 | - | 1 | 177 | ||
| BC090895.1 | - | 1 | 145 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928327| Gene Symbol: LOC101928327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130782.1 | 1 | 172 | |||
| GenBank mRNA | BC021184.1 | - | 1 | 177 | ||
| BC090895.1 | - | 1 | 145 | |||