siRNA Details
| Gene Symbol: LINC00635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015414.1 | 11 | 1376 | |||
| GenBank mRNA | BC045586.1 | - | 11 | 1376 | ||
| DB527077.2 | - | 2 | 270 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00635| Gene Symbol: LINC00635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015414.1 | 11 | 1376 | |||
| GenBank mRNA | BC045586.1 | - | 11 | 1376 | ||
| DB527077.2 | - | 2 | 270 | |||