siRNA Details
Gene Symbol: LINC00635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015414.1 | 8 | 772 | |||
GenBank mRNA | BC045586.1 | - | 8 | 772 | ||
BC104745.1 | - | 4 | 366 | |||
BC104746.1 | - | 3 | 311 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015414.1 | 8 | 772 | |||
GenBank mRNA | BC045586.1 | - | 8 | 772 | ||
BC104745.1 | - | 4 | 366 | |||
BC104746.1 | - | 3 | 311 |