siRNA Details
| Gene Symbol: SHISA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080505.1 | NP_001073974.1 | 2 | 1082 | ||
| GenBank mRNA | BC012029.1 | - | 1 | 582 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SHISA3| Gene Symbol: SHISA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080505.1 | NP_001073974.1 | 2 | 1082 | ||
| GenBank mRNA | BC012029.1 | - | 1 | 582 | ||