siRNA Details
| Gene Symbol: LOC100505841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011543086.2 | XP_011541388.1 | 4 | 287 | ||
| XM_017008912.1 | XP_016864401.1 | 4 | 1190 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505841| Gene Symbol: LOC100505841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011543086.2 | XP_011541388.1 | 4 | 287 | ||
| XM_017008912.1 | XP_016864401.1 | 4 | 1190 | |||