siRNA Details
| Gene Symbol: CYLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021118.2 | NP_066941.1 | 4 | 270 | ||
| XM_005262086.3 | XP_005262143.1 | 4 | 267 | |||
| GenBank mRNA | BC021669.1 | - | 4 | 254 | ||
| BC058019.1 | - | 4 | 254 | |||
| BC070355.1 | - | 4 | 247 | |||
| BC126461.1 | - | 4 | 234 | 8992177 | ||
| DB028115.1 | - | 4 | 267 | |||
| DB049397.1 | - | 4 | 270 | |||
| Z22780.1 | - | 2 | 55 | |||

CYLC1
Genomic Map