siRNA Details
| Gene Symbol: CLEC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080511.2 | NP_001073980.2 | 3 | 334 | ||
| XM_017011770.1 | XP_016867259.1 | 3 | 285 | |||
| GenBank mRNA | AK057548.1 | - | 1 | 1237 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLEC2L| Gene Symbol: CLEC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080511.2 | NP_001073980.2 | 3 | 334 | ||
| XM_017011770.1 | XP_016867259.1 | 3 | 285 | |||
| GenBank mRNA | AK057548.1 | - | 1 | 1237 | ||