siRNA Details
Gene Symbol: LINC00032 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026679.1 | 13 | 2339 | |||
GenBank mRNA | AF418573.2 | - | 13 | 2339 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00032 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026679.1 | 13 | 2339 | |||
GenBank mRNA | AF418573.2 | - | 13 | 2339 |