siRNA Details
| Gene Symbol: PANK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK001644.1 | - | 3 | 410 | ||
| AK223546.1 | - | 3 | 411 | |||
| AK295301.1 | - | 3 | 442 | |||
| AK309860.1 | - | 2 | 953 | |||
| AL442072.1 | - | 3 | 387 | |||
| BC043496.1 | - | 3 | 405 | 5244472 | ||
| DC327227.1 | - | 3 | 442 | |||

PANK4
Genomic Map