siRNA Details
Gene Symbol: CCDC87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018219.2 | NP_060689.2 | 1 | 810 | ||
GenBank mRNA | AK001648.1 | - | 1 | 790 | ||
BC034469.2 | - | 1 | 810 | 4823402 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CCDC87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018219.2 | NP_060689.2 | 1 | 810 | ||
GenBank mRNA | AK001648.1 | - | 1 | 790 | ||
BC034469.2 | - | 1 | 810 | 4823402 |