siRNA Details
| Gene Symbol: LOC441081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073404.1 | 1 | 5137 | |||
| Gene Symbol: LOC101929599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017010105.1 | XP_016865594.1 | 1 | 2395 | ||
| Gene Symbol: LOC107986351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017010104.1 | XP_016865593.1 | 1 | 2380 | ||

LOC441081
Genomic Map