siRNA Details
| Gene Symbol: LINC00858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038220.1 | 6 | 2466 | |||
| GenBank mRNA | AK056561.1 | - | 6 | 2466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00858| Gene Symbol: LINC00858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038220.1 | 6 | 2466 | |||
| GenBank mRNA | AK056561.1 | - | 6 | 2466 | ||