siRNA Details
| Gene Symbol: LINC00466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038252.2 | 5 | 688 | |||
| GenBank mRNA | BC040309.1 | - | 5 | 661 | ||
| DB447804.2 | - | 4 | 603 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00466| Gene Symbol: LINC00466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038252.2 | 5 | 688 | |||
| GenBank mRNA | BC040309.1 | - | 5 | 661 | ||
| DB447804.2 | - | 4 | 603 | |||