siRNA Details
| Gene Symbol: LOC200726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102659.1 | NP_001096129.1 | 2 | 370 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC200726| Gene Symbol: LOC200726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102659.1 | NP_001096129.1 | 2 | 370 | ||