siRNA Details
Gene Symbol: FBXO45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105573.1 | NP_001099043.1 | 3 | 1687 | ||
GenBank mRNA | AK025697.1 | - | 3 | 1413 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FBXO45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105573.1 | NP_001099043.1 | 3 | 1687 | ||
GenBank mRNA | AK025697.1 | - | 3 | 1413 |