siRNA Details
| Gene Symbol: LYRM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076680.1 | NP_001070148.1 | 2, 3 | 215 | ||
| XM_011524491.2 | XP_011522793.1 | 2, 3 | 414 | |||
| XM_017024325.1 | XP_016879814.1 | 2, 3 | 398 | |||
| GenBank mRNA | AK055808.1 | - | 2, 3 | 191 | ||
| AY251535.1 | - | 1, 2 | 84 | |||
| BC018092.2 | - | 1, 2 | 148 | |||
| DB474433.1 | - | 2, 3 | 215 | |||
| DR155808.1 | - | 2, 3 | 180 | |||

LYRM9
Genomic Map