siRNA Details
| Gene Symbol: ZBTB7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039360.2 | NP_001034449.1 | 1 | 175 | ||
| GenBank mRNA | AK296709.1 | - | 1 | 175 | ||
| AM988836.1 | - | 1 | 163 | |||
| EF120357.1 | - | 1 | 227 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZBTB7C| Gene Symbol: ZBTB7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039360.2 | NP_001034449.1 | 1 | 175 | ||
| GenBank mRNA | AK296709.1 | - | 1 | 175 | ||
| AM988836.1 | - | 1 | 163 | |||
| EF120357.1 | - | 1 | 227 | |||