siRNA Details
| Gene Symbol: CTAGE10P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003268.3 | 1 | 275 | |||
| GenBank mRNA | BC030655.1 | - | 1 | 174 | ||
| BI463404.1 | - | 1 | 172 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTAGE10P| Gene Symbol: CTAGE10P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003268.3 | 1 | 275 | |||
| GenBank mRNA | BC030655.1 | - | 1 | 174 | ||
| BI463404.1 | - | 1 | 172 | |||