siRNA Details
| Gene Symbol: WAC-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033805.1 | 3 | 1265 | |||
| GenBank mRNA | AK001143.1 | - | 1 | 476 | ||
| BC045818.1 | - | 3 | 1245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WAC-AS1| Gene Symbol: WAC-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033805.1 | 3 | 1265 | |||
| GenBank mRNA | AK001143.1 | - | 1 | 476 | ||
| BC045818.1 | - | 3 | 1245 | |||