siRNA Details
| Gene Symbol: PRSS54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080492.1 | NP_001073961.1 | 5 | 760 | ||
| NM_001305173.1 | NP_001292102.1 | 5 | 757 | |||
| NM_001305174.1 | NP_001292103.1 | 4 | 582 | |||
| GenBank mRNA | AK058068.1 | - | 5 | 702 | ||
| AK302175.1 | - | 4 | 512 | |||
| AL137484.1 | - | 1 | 50 | |||
| BC057843.1 | - | 5 | 749 | 5296090 | ||

PRSS54
Genomic Map