siRNA Details
| Gene Symbol: CCDC162P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC128118.1 | - | 2 | 222 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC162P| Gene Symbol: CCDC162P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC128118.1 | - | 2 | 222 | ||