siRNA Details
| Gene Symbol: ENDOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015036.2 | NP_055851.1 | 2 | 1902 | ||
| GenBank mRNA | AB020637.1 | - | 2 | 1853 | ||
| AK026560.1 | - | 1 | 1415 | |||
| BC026191.1 | - | 2 | 1869 | 4816880 | ||
| BC071171.1 | - | 2 | 1853 | 5755016 | ||

ENDOD1
Genomic Map